Magnetic hotplate stirrer, RCT basic
Leverandør: IKA
25005927
442-1314EA 12925 NOK
442-1314
Magnetic hotplate stirrer, RCT basic
Omrørere Magnetrørere med varmeplate
The RCT basic magnetic stirrer provides reliability, an exceptionally long product lifetime and the highest safety standards. RCT basic is suitable for stirring tasks up to 20 L (H₂O) and reaches temperatures up to 310 °C. With the connection option for an external temperature sensor (PT1000.60), the temperature can be measured and controlled directly in the reaction medium. Thanks to insulation of the aluminium heating plate, maintenance-free EC motor and electronic switching power supply, RCT basic features excellent energy efficiency as well as reduced self-heating of the heating plateMore Product Information
Bestill nå
SPESIFIKASJONER
Modell RCT basic
Plate material Aluminium legering
Environmentally Preferable
Platemål Ø 135 mm
Max. stirring capacity H₂O 20 L
Speed range 50 - 1500 min⁻¹
B×D×H 160×85×270 mm
Temperaturkontrollnøyaktighet ±0,5 (external sensor (via ETS-D5))
Temperaturområde Ambient +13…310 °C
Temperaturinnstilling nøyaktighet ±5 K
Varmeeffekt 600 W
Vekt 2,4 kg