Digital multi-tube vortex, MULTI-TX5
Leverandør: VELP SCIENTIFIC
F202A0460
442-1409EA 29525 NOK
442-1409
Digital multi-tube vortex, MULTI-TX5
Risteapparater Reagensrørrister
The MULTI-TX5 Digital is the ideal digital multi-tube vortex mixer for high sample throughput. The innovative and compact design ensures maximum operating comfort for mixing applications requiring different test tubes.
  • Stirring speed from 100 up to 2500 m⁻¹ thanks to the powerful brushless motor
  • Designed for outstanding comfort, maximum resistance, and minimum footprint
  • Easy to operate digital interface with icons
  • Built-in timer allows programming of unsupervised operation
  • Programmable auto-reverse of stirring direction
  • Pulse Mode enables cyclical vigorous