Immersion thermostats, EH 4 basic
Leverandør: IKA
3164000 3164002
442-2393EA 0 NOK
442-2393 IKAA3164002
Immersion thermostats, EH 4 basic
Termostatbad Termostatbad, åpent Termostatkontrollere
Immersion circulator for temperature control of up to 100 °C in open baths (with depth of at least 150 mm)
  • Complies with all safety requirements for electrically operated devices
  • Intended for supervised use
  • Operate with non-flammable liquids only
  • Suitable for all standard bath vessels
Leveringsinformasjon: Supplied with universal clamp.