Dodecyltrimethylammonium hydrogen sulphate ≥99.0% (by titrimetric analysis), LiChropur™ for ion pair chromatography, Supelco®
Leverandør: Merck
Warning
M-Clarity™ quality level = MQ100
44243-10G
44243-10GEA 7050 NOK
44243-10G
Dodecyltrimethylammonium hydrogen sulphate ≥99.0% (by titrimetric analysis), LiChropur™ for ion pair chromatography, Supelco®
Dodecyltrimethylammonium hydrogen sulphate
Formel: CH₃(CH₂)₁₁N(CH₃)₃HSO₄
MDL nummer: MFCD01863032
CAS-nummer: 103999-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba356d0fedd3cb87bcac0014c4a802547f2ea04d&vid=ee7d661fe0c0fb223f18709e5d6efc96bfb5efca&prid=web&clid=SAPDAM