Pesticide reference standard, Endrin aldehyde, analytical standard, Supelco®

Leverandør: Merck

442578
442578.EA 1250 NOK
442578.
Pesticide reference standard, Endrin aldehyde, analytical standard, Supelco®
Standarder Organiske standarder

Bestill nå

SPESIFIKASJONER

Beskrivelse Endrin aldehyde
Environmentally Preferable
CAS-nr 7421-93-4
Formel C₁₂H₈Cl₆O
Lagringstemp RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR