Magnesiumklorid heksahydrat, Millipore®

Leverandør: Merck Millipore Calbiochem

442615-500
442615-500EA 1475 NOK
442615-500
Magnesiumklorid heksahydrat, Millipore®
Magnesiumklorid heksahydrat
Formel: MgCl₂·6H₂O
Kokepunkt: 1412 °C (1013 hPa)
Smeltepunkt: 117 °C
Tetthet: 1,57 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149781
CAS-nummer: 7791-18-6
EINECS: 232-094-6
Merck Indeks: 13,05684

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR