Formate standard for ion chromatography, TraceCERT®, Supelco®
Leverandør: Merck
44293-100ML
44293-100MLEA 615 NOK
44293-100ML
Formate standard for ion chromatography, TraceCERT®, Supelco®
Standarder Anionstandarder Formate Standards
Prepared with calcium formate and water, filtered through a 0,2 μm membrane filter and stabilised with sodium azide.
Sertifiseringer: Certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034.