Aluminium ≥99.9999% (metals basis), folie, Puratronic®, annealed, Thickness: 0.5 mm (0.02 in)

Leverandør: Thermo Fisher Scientific

44332.FI 44332.GH 44332.HP
44332.FIEA 1725 NOK
44332.FI 44332.GH 44332.HP
Aluminium ≥99.9999% (metals basis), folie, Puratronic®, annealed, Thickness: 0.5 mm (0.02 in)
Aluminium

Purity: 99.9999% CAS No.: 7429-90-5 Physical Form: ≈3.4g/50x50mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029

Formel: Al
Kokepunkt: 2327 °C (1013 hPa)
Smeltepunkt: 660 °C
Tetthet: 2,7 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00134029
CAS-nummer: 7429-90-5
EINECS: 231-072-3
Merck Indeks: 13,00321

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR