Tris-(hydroksymetyl)aminometan ≥99%, Electran for electrophoresis
Leverandør: VWR Chemicals
Warning
443866GEA 4200 NOK
443866G
Tris-(hydroksymetyl)aminometan ≥99%, Electran for electrophoresis
Tris-(hydroksymetyl)aminometan
Formel: NH₂C(CH₂OH)₃
Kokepunkt: 357 °C (1013 hPa)
Smeltepunkt: 169…172 °C
Tetthet: 1,328 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004679
CAS-nummer: 77-86-1
EINECS: 201-064-4
Merck Indeks: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Assay Min. 99.00%
Appearance White or colorless crystal powder
Identity (IR) Passes test
DNases Not detected
RNases Not detected
Proteases Not detected
Absorbance (260 nm) (10%) Max. 0.030
Absorbance (280 nm) (10%) Max. 0.020
pH (20 °C; 1 mol/l) 10.50 - 11.50
Heavy metals (as Pb) Max. 0.005%
Water Max. 0.3%
Cu (Copper) Max. 0.0001%
Fe (Iron) Max. 0.0001%
Pb (Lead) Max. 0.0001%