Platform rocker, PMR-30
Leverandør: GRANT INSTRUMENTS
PMR-30
444-0341EA 7850 NOK
444-0341
Platform rocker, PMR-30
Risteapparater Risteapparater, roterende/gyngende
The compact and highly functional PMR-30 variable speed, fixed angle platform rocker is ideally suited for personal use, it provides a smooth side to side rocking motion for gentle sample agitation in tubes, culture flasks, dishes and boxes. Applications include gel staining, destaining, antibody staining, washes, hybridisations, and blots. The reliable motor with direct drive mechanism produces regulated and reproducible rocking throughout the speed range.
  • Continuous or timer operation; integral digital timer 1 min to 24 h, with automatic switch-off, time