VWR® Advanced Magnetic Stirrers
Supplier: VWR Collection
444-0567EA 4925 NOK
444-0567 444-0568 444-0570 444-0571 444-0572 444-0573 444-0574
VWR® Advanced Magnetic Stirrers
Stirrers Magnetic Stirrers
VWR® Advanced magnetic stirrers deliver accurate and repeatable results. These easy to use microprocessor-controlled units have an analogue dial with rpm adjustment markings. Rear housing features an integral support rod holder with locking knob to accept an optional support rod and clamp kit.
  • Built-in support rod holder
  • Microprocessor control with analogue dial
  • Chemically resistant housing
  • Ceramic top plate
The low profile design takes up less space and fits into fume hoods. Spill resistant housing channels fluids away from internal components.More Product Information
Order Now