Shakers, Roller 6 and 10 basic and digital
Leverandør: IKA
4010000 4012000 4011000 4013000
444-0819EA 23225 NOK
444-0819 444-0821 444-0820 444-0822
Shakers, Roller 6 and 10 basic and digital
Risteapparater Risteapparater, roterende/gyngende
Roller shakers with 6 or 10 rolls, provide smooth rocking and rolling action, basic models have fixed speed, and the digital units feature infinitely adjustable speed. Used for mixing blood samples, solid and liquid suspensions and viscous samples. The unit can be used with all conventional tubes and cylindrical bottles. Easily removable attachments enable quick cleaning in case of sample spills.
  • Can be used at 4 to 50 °C in an incubator or cold room
  • Suitable for continuous operation
  • Removable rolls enable shaking of larger tubes
  • Easy operation via touch