VWR®, Multi Purpose Clamps, 3-Prong
Leverandør: VWR Collection
444-0916EA 820 NOK
444-0916
VWR®, Multi Purpose Clamps, 3-Prong
Klemmer Universalklemmer
The multi purpose clamp is designed to securely hold every type of laboratory glassware and apparatus.
  • Large grip adjustment range
  • Three prong construction
  • Dual adjust
Long, seamless nickel-plated brass tubing attaches clamp head securely and offers easy positioning in the deepest fume hoods. Clamps are constructed with round extension arms, which allow the clamps to be rotated 360°.
Extension arms also allow placement of apparatus at various distances from lab-frames without compromising the integrity of the experiment.
Bestillingsinformasjon: More Product Information