VWR®, Support Stand, Rectangular Base, Cast Iron
Leverandør: VWR Collection
444-0918EA 2225 NOK
444-0918
VWR®, Support Stand, Rectangular Base, Cast Iron
Stativer
Cast iron support stand bases are constructed with black enamel finish for chemical resistance and durability.
It features a built-in support rod holder with locking knob that accepts a 13 mm diameter support rod.
Bestillingsinformasjon: Support rods are not included.