Overhead shakers, Trayster basic and digital
Leverandør: IKA
4005000 4006000
444-1037EA 13775 NOK
444-1037 444-1038
Overhead shakers, Trayster basic and digital
Risteapparater Risteapparater, frem og tilbake bevegelse
Overhead shakers with vertical rotation for smooth yet effective mixing, the basic model has fixed speed, and the digital unit features infinitely adjustable speed. Ideal for biological samples, particularly blood. Suitable for separating and mixing processes of powdered and liquid samples in tubes up to 50 ml. Easily removable attachments provide for quick cleaning in case of sample spills. Different attachments enable units to be used for a variety of applications.
  • Can be used at 4 to 50 °C in an incubator or cold room
  • Easy operation via touch keypad