Reciprocating shakers, Promax
Leverandør: Heidolph Instruments GmbH & Co.KG
543-22332-00 542-20020-05 543-22332-06 542-20020-02 543-22332-02
444-1320EA 26600 NOK
444-1320 444-1255 444-1256 444-1473 444-1478
Reciprocating shakers, Promax
Risteapparater Risteapparater, sideveis
Promax shakers have a one dimensional back and forth motion for vigorous mixing of flasks and separating funnels in chemical applications, using analogue dial controls for reciprocating frequency. The encapsulated housing ensures virtually no heat transfer from motor to platform and a sealed drive is isolated from spills.
  • Speed continuously adjustable
  • Modular system; combine Promax 1020 with Incubator 1000 with choice of a low or high incubator hood
  • Process timer with audible signal allows for unattended operation
  • Promax 1020; digital timer up to 999 min,