Mangan (II) klorid tetrahydrat ≥99.99% (metals basis)

Leverandør: Thermo Fisher Scientific
Danger

44442.06 44442.14 44442.22
44442.06EA 655 NOK
44442.06 44442.14 44442.22
Mangan (II) klorid tetrahydrat ≥99.99% (metals basis)
Mangan (II) klorid tetrahydrat

Purity: 99.99% CAS No.: 13446-34-9 Molecular Formula: MnCl2.4H2O Formula Weight: 197.90 (125.84anhy) Physical Form: Crystalline Density: 2.010 Melting Point: 58° Merck Reference: 14,5728 MDL No.: MFCD00149792

Formel: MnCl₂·4H₂O
Kokepunkt: 1190 °C (1013 hPa)
Smeltepunkt: 58 °C
Tetthet: 2,010
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149792
CAS-nummer: 13446-34-9
EINECS: 231-869-6
FN: 3077
ADR: 9,III
Merck Indeks: 14,5728

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR