Cadmium(II) bromide ≥99.999% (metals basis)

Leverandør: Thermo Scientific
Warning

Synonymer: Cadmium bromide

44498.22
44498.22EA 3300 NOK
44498.22
Cadmium(II) bromide ≥99.999% (metals basis)
Cadmium(II) bromide

Purity: 99.999% CAS No.: 7789-42-6 Molecular Formula: CdBr2.xH2O Formula Weight: 272.22(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White to pale yellow Odour: Odourless UN No.: 2570 MDL No.: MFCD00150204

Formel: Br₂Cd
MW: 272,22 g/mol
Kokepunkt: 863 °C (1013 hPa)
Smeltepunkt: 567 °C
Tetthet: 5,19 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00015996
CAS-nummer: 7789-42-6
EINECS: 232-165-1
FN: 2570
ADR: 6.1,III
Merck Indeks: 12,01651

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR