Triisopropylsilane 98%, AcroSeal®

Leverandør: Thermo Fisher Scientific
Warning

446171000
446171000.EA 2025 NOK
446171000.
Triisopropylsilane 98%, AcroSeal®
Triisopropylsilane
Formel: C₉H₂₂Si
Kokepunkt: 84…86 °C (35 mmHg)
Tetthet: 0,773 g/cm³
Flash punkt: 37 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009657
CAS-nummer: 6485-79-6
FN: 1993
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4360 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR