J.T.Baker® BAKER-flex®, Precoated flexible TLC sheet
Leverandør: Avantor
4463-04EA 8125 NOK
4463-04
J.T.Baker® BAKER-flex®, Precoated flexible TLC sheet
TLC-plater
Ready-to-use in thin layer chromatography applications, Baker-flex® precoated flexible plastic sheet have a hard, abrasion-resistant coating and solvent-resistant substrate for rapid, clean separations.
The sheets can be written on and cut to size. They are available with or without fluorescent indicator.