Tellur 1000 mg/L Te in 5% salpetersyre (fra Te) Specpure® enkeltgrunnstoffsstandard til ICP
Leverandør: Thermo Fisher Scientific
44632.AD 44632.AE 44632.AP
44632.ADEA 1200 NOK
44632.AD 44632.AE 44632.AP
Tellur 1000 mg/L Te in 5% salpetersyre (fra Te) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Tellurium Standards
Molecular Formula: Te in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134062
Sertifiseringer: Traceable to NIST.