Tellur 1000 mg/L Te in 5% salpetersyre (fra Te) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

44632.AD 44632.AE 44632.AP
44632.ADEA 1200 NOK
44632.AD 44632.AE 44632.AP
Tellur 1000 mg/L Te in 5% salpetersyre (fra Te) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Tellurium Standards

Molecular Formula: Te in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134062


Sertifiseringer: Traceable to NIST.

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00134062

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR