Keto Reagent Kit, Amplifex™, Supelco®
Leverandør: Merck
4465962-1KT
4465962-1KTEA 6090 NOK
4465962-1KT
Keto Reagent Kit, Amplifex™, Supelco®
Keto Reagent Kit
It was used in targeted metabolomic analysis, in a study done to understand the mitochondrial thiol modification, by targeted electrophile, which inhibits metabolism in breast adenocarcinoma cells. It may also be used in the oxosteroid chemical derivatization kit, in a study to determine the neurosteroids in sea lamprey by UPLC-MS/MS.
Leveringsinformasjon: Kit contains a protocol, 2 tubes of Amplifex™ Keto reagent and 2 tubes of diluent. Each tube contains enough reagent to complete 25 reactions.
NB: Amplifex is a trademark of AB Sciex Pte. Ltd.