D-Biotin-N-succinimidyl ester ≥98%

Leverandør: Thermo Fisher Scientific

44771.01 44771.03 44771.MF
44771.01EA 4050 NOK
44771.01 44771.03 44771.MF
D-Biotin-N-succinimidyl ester ≥98%
D-Biotin-N-succinimidyl ester

Purity: 98% CAS No.: 35013-72-0 Molecular Formula: C14H19N3O5S Formula Weight: 341.40 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 212-215° Beilstein Reference: 4720781 MDL No.: MFCD00078531


Applications: In coupling reactions to amino acids, peptides and proteins through primary amines

Formel: C₁₄H₁₉N₃O₅S
Smeltepunkt: 212…215 °C
MDL nummer: MFCD00078531
CAS-nummer: 35013-72-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR