Instrument calibration(ICAL) standard, Solution A, Specpure®

Leverandør: Thermo Fisher Scientific

44855.AEEA 2525 NOK
44855.AE
Instrument calibration(ICAL) standard, Solution A, Specpure®
Standarder Kationstandarder Multi-Element Standards

Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD08064526


Concentration:
Ag 200 µg/ml ±2%
Ba 1,000 µg/ml ±2%
Be 400 µg/ml ±2%
Cd 500 µg/ml ±2%
Co 1,000 µg/ml ±2%
Cu 1,000 µg/ml ±2%
Fe 1,000 µg/ml ±2%
Mn 1,000 µg/ml ±2%
Ni 1,000 µg/ml ±2%
Pb 1,000 µg/ml ±2%
Tl 1,000 µg/ml ±2%
Zn 1,000 µg/ml ±2%

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD08064526

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR