Saltsyre 6 mol/l (6 N)

Leverandør: Thermo Fisher Scientific
Danger

44921.K2
44921.K2EA 780 NOK
44921.K2
Saltsyre 6 mol/l (6 N)
Saltsyre
Formel: HCl
Tetthet: 1,06 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8,II

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR