Saltsyre 37%, ARISTAR® for trace analysis

Leverandør: VWR Chemicals
Danger

Synonymer: Hydrochloric acid solution, Muriatic acid

450021YEA 1750 NOK
450021Y 450027X
Saltsyre 37%, ARISTAR® for trace analysis
Saltsyre
Formel: HCl
MW: 36.46 g/mol
Smeltepunkt: –30 °C
Tetthet: 1.18 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8 II
REACH: 01-2119484862-27

Bestill nå

Spesifikasjon Testresultater

Assay 36.0 - 38.0 %
Colour Max. 10 APHA
Br (Bromide) Max. 50 ppm
Free chlorine Max. 0.5 ppm
PO4 (Phosphate) Max. 0.1 ppm
SO4 (Sulphate) Max. 0.5 ppm
SO3 (Sulphite) Max. 0.500 ppm
Ag (Silver) Max. 0.010 ppm
Al (Aluminium) Max. 0.050 ppm
As (Arsenic) Max. 0.010 ppm
Au (Gold) Max. 0.020 ppm
B (Boron) Max. 0.010 ppm
Ba (Barium) Max. 0.005 ppm
Be (Beryllium) Max. 0.010 ppm
Bi (Bismuth) Max. 0.020 ppm
Ca (Calcium) Max. 0.100 ppm
Cd (Cadmium) Max. 0.003 ppm
Co (Cobalt) Max. 0.005 ppm
Cr (Chromium) Max. 0.005 ppm
Cu (Copper) Max. 0.002 ppm
Fe (Iron) Max. 0.050 ppm
Ga (Gallium) Max. 0.020 ppm
Hg (Mercury) Max. 0.001 ppm
In (Indium) Max. 0.010 ppm
K (Potassium) Max. 0.010 ppm
Li (Lithium) Max. 0.010 ppm
Mg (Magnesium) Max. 0.020 ppm
Mn (Manganese) Max. 0.005 ppm
Mo (Molybdenum) Max. 0.005 ppm
NH4 (Ammonium) Max. 1 ppm
Na (Sodium) Max. 0.050 ppm
Ni (Nickel) Max. 0.010 ppm
Pb (Lead) Max. 0.002 ppm
Pt (Platinum) Max. 0.010 ppm
Sn (Tin) Max. 0.010 ppm
Sr (Strontium) Max. 0.005 ppm
Ti (Titanium) Max. 0.010 ppm
Tl (Thallium) Max. 0.010 ppm
V (Vanadium) Max. 0.0050 ppm
Zn (Zinc) Max. 0.010 ppm
Zr (Zirconium) Max. 0.005 ppm
Ignition residue (SO4) Max. 2.0 ppm
Residue on evaporation Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR