Bunsen burner, cartridge type
Leverandør: USBECK Laborgeräte
1422
451-0156EA 1200 NOK
451-0156
Bunsen burner, cartridge type
Termostater Gassbrennere
Nickel plated brass Bunsen burner ideal for heating or sterilising instruments and laboratory glassware.
  • With air regulation needle valve
Height: 85 mm
Bestillingsinformasjon: Supplied without cartridge, please order cartridge separately.