Magnesium, cubes ∼ 19 mm (0.75 in) each side, Length: 19mm (0.75in)
Leverandør: Thermo Fisher Scientific
45119.KF
45119.KFEA 1150 NOK
45119.KF
Magnesium, cubes ∼ 19 mm (0.75 in) each side, Length: 19mm (0.75in)
Magnesium
CAS No.: 7439-95-4 Physical Form: ≈10-15g/cube Appearance: Silver-Coloured Odour: Odourless MDL No.: MFCD00085308
Formel: Mg
Kokepunkt: 1107 °C (1013 hPa)
Smeltepunkt: 651 °C
Tetthet: 1,74 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00085308
CAS-nummer: 7439-95-4
EINECS: 231-104-6
FN: 1869
ADR: 4.1,III
Merck Indeks: 13,05675