Beryllium 10 mg/L Be in 5% salpetersyre (fra BeO.(Be(OAc)₂)₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

45248.AEEA 940 NOK
45248.AE
Beryllium 10 mg/L Be in 5% salpetersyre (fra BeO.(Be(OAc)₂)₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Beryllium Standards

Molecular Formula: Be₄O(C₂H₃O₂)6 in 2% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134032


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00134032

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR