Krom 10 mg/L Cr in 5% salpetersyre (fra Cr(NO₃)₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

45251.AEEA 1250 NOK
45251.AE
Krom 10 mg/L Cr in 5% salpetersyre (fra Cr(NO₃)₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Chromium Standards

Molecular Formula: Cr(NO₃)₃ in 2% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010944


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00010944

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR