Kobalt 10 mg/L Co in 5% salpetersyre (fra Co) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

45252.AEEA 1200 NOK
45252.AE
Kobalt 10 mg/L Co in 5% salpetersyre (fra Co) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Cobalt Standards

Molecular Formula: Co in 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00010935


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00010935

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR