Germanium 10 mg/L Ge in 5% salpetersyre with hydrofluoric acid (max. 1%) (de Ge) Specpure® standard til AAS
Leverandør: Thermo Fisher Scientific
45254.AEEA 1350 NOK
45254.AE
Germanium 10 mg/L Ge in 5% salpetersyre with hydrofluoric acid (max. 1%) (de Ge) Specpure® standard til AAS
Standarder Kationstandarder Germanium Standards
Sertifiseringer: Traceable to NIST