Supelco® PESTANAL® Organic Reference Standard, Cyprofuram

Leverandør: Merck

45413-100MG
45413-100MGEA 530 NOK
45413-100MG
Supelco® PESTANAL® Organic Reference Standard, Cyprofuram
Standarder Organiske standarder

  • GHS signal: GHS06, GHS09


NB: P264 - P270 - P273 - P280 - P301 + P310 - P302 + P352 + P312

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 69581-33-5
MDL No. MFCD00078634
Molekylvekt 279,72
Hardhetsklasse beskrivelse H301 - H312 - H410
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR