Difenylamin, Supelco®

Leverandør: Merck
Danger

Synonymer: N-Phenylaniline, N-Phenylbenzeneamine

M-Clarity™ quality level = MQ100

45456-250MG
45456-250MGEA 294 NOK
45456-250MG
Difenylamin, Supelco®
Difenylamin
Formel: (C₆H₅)₂NH
MW: 169.23 g/mol
Kokepunkt: 302
Smeltepunkt: 53…54 °C
Tetthet: 1.165 g/cm³ (20 °C)
Flash punkt: 153 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003014
CAS-nummer: 122-39-4
EINECS: 204-539-4
FN: 3077
ADR: 9 III
Merck Indeks: 13,03349
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0977543f74dc547c2762b27ae65ecb5903e87394&vid=c81475ff1bf619037e4b83948e285cba701e6b26&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR