Supelco® PESTANAL® Organic Reference Standard, Dodemorph

Leverandør: Merck

45465-250MG
45465-250MGEA 900 NOK
45465-250MG
Supelco® PESTANAL® Organic Reference Standard, Dodemorph
Standarder Organiske standarder

  • GHS signal: GHS07, GHS09


NB: P260 - P273 - P280 - P303 + P361 + P353 - P304 + P340 + P310 - P305 + P351 + P338

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05163992541172fd3dcaada57b0775845f17770d&vid=09c994d9b2df725804f191e0edf6eea7c846c2d4&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) 4-Cyclododecyl-2,6-dimethylmorpholine
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 1593-77-7
MDL No. MFCD00155442
Formel C₁₈H₃₅NO
Molekylvekt 281,48
Hardhetsklasse beskrivelse H314 - H317 - H361d - H373 - H410
Flammepunkt Not applicable
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR