Iron-chrome based high temperature gas shift catalyst, HiFUEL® W210
Leverandør: Thermo Fisher Scientific
Danger
45467.A1 45467.36 45467.A4
45467.A1EA 2750 NOK
45467.A1 45467.36 45467.A4
Iron-chrome based high temperature gas shift catalyst, HiFUEL® W210
Iron-chrome based high temperature gas shift catalyst
Physical Form: Pellets, 5.4mm x 3.6mm Appearance: Red-brown