Tube furnaces with stand for horizontal or vertical operation, RT
Leverandør: NABERTHERM
RT332LAN RT332OAN RT512LAN RT512OAN RT522LAN RT522OAN
455-2098EA 0 NOK
455-2098 455-2099 455-2103 455-2104 455-2105 455-2106
Tube furnaces with stand for horizontal or vertical operation, RT
Ovner Rørovner
These compact tube furnaces are used when laboratory experiments must be performed horizontally, vertically, or at specific angles. The ability to configure the angle of tilt and the working height, make these tube furnaces suitable for integration into existing process systems.
  • Compact design with integrated control system in furnace base
  • Choice of models with Tmax 1100, 1300 or 1500 °C
  • Only fibre materials which are not classified as carcinogenic according to TRGS 905, class 1 or 2 are used
  • Vertical or horizontal operation freely adjustable, working