Aluminium in fortynnet salpetersyre (fra Al(NO3)3.9H2O) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455002CEA 945 NOK
455002C 455012E 455014G 455004E
Aluminium in fortynnet salpetersyre (fra Al(NO3)3.9H2O) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Aluminium Standards
Al(NO₃)₃.9H₂O in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with Certificate of Analysis.