Antimon enkeltgrunnstoffsstandarder til ICP, ARISTAR®
Leverandør: VWR Chemicals
455022GEA 1550 NOK
455022G 455024Y 455034K
Antimon enkeltgrunnstoffsstandarder til ICP, ARISTAR®
Standarder Kationstandarder Antimony Standards
Sb in HNO₃ 2% tr. tartaric acid.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.