Arsen 1.000 mg/L i fortynnet salpetersyre (fra As) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455042KEA 1020 NOK
455042K 455044M
Arsen 1.000 mg/L i fortynnet salpetersyre (fra As) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Arsenic Standards
As in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.