Arsen 10.000 mg/L i fortynnet salpetersyre (fra As) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455054XEA 4000 NOK
455054X 455052M
Arsen 10.000 mg/L i fortynnet salpetersyre (fra As) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Arsenic Standards
As in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.