Barium 10.000 mg/L i fortynnet salpetersyre (fra Ba(NO3)2) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455072QEA 2075 NOK
455072Q
Barium 10.000 mg/L i fortynnet salpetersyre (fra Ba(NO3)2) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Barium Standards
Ba(NO₃)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.