Beryllium 1.000 mg/l i fortynnet salpetersyre, ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455082SEA 2075 NOK
455082S 455084U
Beryllium 1.000 mg/l i fortynnet salpetersyre, ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Beryllium Standards
BeO.(Be(OAc)₂)₃) in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.