Bismut 10.000 mg/l i fortynnet salpetersyre (fra Bi) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455112HEA 2125 NOK
455112H 455114J
Bismut 10.000 mg/l i fortynnet salpetersyre (fra Bi) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Bismuth Standards
Bi in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.