Kadmium 10.000 mg/l i fortynnet salpetersyre (fra Cd) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455154REA 5825 NOK
455154R 455152P
Kadmium 10.000 mg/l i fortynnet salpetersyre (fra Cd) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Cadmium Standards
Cd in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.