Kalsium 1.000 mg/l i fortynnet salpetersyre (fra CaCO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455162REA 1080 NOK
455162R 455164T
Kalsium 1.000 mg/l i fortynnet salpetersyre (fra CaCO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Calcium Standards
CaCO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with Certificate of Analysis.