Cerium 10.000 mg/l i fortynnet salpetersyre (fra (NH4)2Ce(NO3)6) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455192AEA 3025 NOK
455192A
Cerium 10.000 mg/l i fortynnet salpetersyre (fra (NH4)2Ce(NO3)6) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Cerium Standards
(NH₄)₂ Ce(NO₃)₆ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.