Cesium 1.000 mg/l i fortynnet salpetersyre (fra CsNO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455204KEA 4400 NOK
455204K 455202Y
Cesium 1.000 mg/l i fortynnet salpetersyre (fra CsNO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Cesium Standards
CsNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.