Krom standardløsninger, standarder for ICP, ARISTAR®
Leverandør: VWR Chemicals
455242QEA 1050 NOK
455242Q 455254U 455252S 455244S 85762.180 85762.270
Krom standardløsninger, standarder for ICP, ARISTAR®
Standarder Kationstandarder Chromium Standards
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.